Ethylmagnesium bromide, 0.9M solution in THF, AcroSeal™
Ethylmagnesium bromide, 0.9M solution in THF, AcroSeal™
Ethylmagnesium bromide, 0.9M solution in THF, AcroSeal™
Ethylmagnesium bromide, 0.9M solution in THF, AcroSeal™
Thermo Scientific Chemicals
Ethylmagnesium bromide, 0.9M solution in THF, AcroSeal™
CAS: 925-90-6 | C2H5BrMg | 133.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA210471000
Price (MYR)
467.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
467.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthylmagnesium bromide
ColorBlack-Brown to Brown
Name Note1M Solution in THF
CAS925-90-6
Health Hazard 1Danger