Metronidazole, 99%
Metronidazole, 99%
Metronidazole, 99%
Thermo Scientific Chemicals
Metronidazole, 99%
Metronidazole, CAS# 443-48-1, is a member of the class of imidazoles with limited antibiotic activity that acts against protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria. | CAS: 443-48-1 | C6H9N3O3 | 171.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number FSA210345000
Price (MYR)
5,857.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
5,857.00
EA
Add to cart
Specifications
Chemical Name or MaterialMetronidazole
CAS443-48-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
General Information
• Metronidazole is a synthetic nitroimidazole derivative with antiprotozoal and antibacterial activities and acts against protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria
• Metronidazole is considered a prodrug that produces nitroso-containing intermediates in the cell that can covalently bind DNA, disrupting helical structure, inducing DNA strand breaks, and inhibiting bacterial nucleic acid synthesis, that results in bacterial cell death
Applications
• Metronidazole shows anti-parasitic and anti-amoebic activities.
• It has been used to prevent the growth of gram-negative anaerobes belonging to the Bacteroides and Fusobacterium, and gram-positive anaerobes such as Peptostreptococcus and Clostridia. This compound affects a high percentage of gram-negative bacteria and has higher tissue penetration that gives it an advantage compared to other compounds.
• This compound was used initially to suppress protozoans such as Entamoeba histolytica, Giardia lamblia, and Trichomonas vaginalis.
• It has been commonly applied for the eradication of Helicobacter pylori.
• It also has a radio-sensitizing effect on hypoxic tumor cells and can have a mutagenic activity in in vivo animal models.
RUO – Research Use Only
General References:
  1. An, B.; Moon, B.; Kim, H. et al. Antibiotic Resistance in Helicobacter pylori Strains and its Effect on H. pylori Eradication Rates in a Single Center in Korea. Annals of Laboratory Medicine 2013, 33, (6), 415-419.
  2. Roach, P.; Wallis, P.; Olson, M. The use of metronidazole, tinidazole and dimetridazole in eliminating trichomonads from laboratory mice. Lab Anim. 1988, 22, (4), 361-364.
  3. Rutgeerts, P.; Hiele, M.; Geboes, K.; Peeters, M.; Penninckx, F.; Aerts, R.; Kerremans, R. Controlled trial of metronidazole treatment for prevention of crohn's recurrence after ileal resection. Gastroenterology. 1995, 108 (6),1617-1621.
  4. Teasley, D. G.; Olson, M. M.; Gebhard, R. L.; Gerding, D. N.; Peterson, L. R.; Schwartz, M. J.; Lee JR, J. T. Prospective Randomised Trial of Metronidazole Versus Vancomycin for Clostridium-Difficile-Associated Diarrhoea and Colitis. The Lancet. 1983, 322 (8358),1043-1046.