(-)-Camphanic acid chloride, 98%
(-)-Camphanic acid chloride, 98%
(-)-Camphanic acid chloride, 98%
(-)-Camphanic acid chloride, 98%
Thermo Scientific Chemicals
(-)-Camphanic acid chloride, 98%
CAS: 39637-74-6 | C10H13ClO3 | 216.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA209890250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material(-)-Camphanic acid chloride
Name Note98%
CAS39637-74-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
View more
Chiral derivatization reagent
RUO – Research Use Only