Benzaldehyde dimethyl acetal, 98%
Benzaldehyde dimethyl acetal, 98%
Benzaldehyde dimethyl acetal, 98%
Benzaldehyde dimethyl acetal, 98%
Thermo Scientific Chemicals
Benzaldehyde dimethyl acetal, 98%
CAS: 1125-88-8 | C9H12O2 | 152.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA209792500
Price (MYR)
805.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
805.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzaldehyde dimethyl acetal
Name Note99+%
CAS1125-88-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning