Hydrazine hydrate, 80% (Hydrazine, 51%), Thermo Scientific Chemicals
Hydrazine hydrate, 80% (Hydrazine, 51%), Thermo Scientific Chemicals
Hydrazine hydrate, 80% (Hydrazine, 51%), Thermo Scientific Chemicals
Hydrazine hydrate, 80% (Hydrazine, 51%), Thermo Scientific Chemicals
Hydrazine hydrate, 80% (Hydrazine, 51%), Thermo Scientific Chemicals
Hydrazine hydrate, 80% (Hydrazine, 51%), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrazine hydrate, 80% (Hydrazine, 51%), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA209592500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point117.2°C
Chemical Name or MaterialHydrazine hydrate
Melting Point-57.0°C
CAS10217-52-4
ColorColorless
View more
Reduction reagent
RUO – Research Use Only