Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™
Thermo Scientific Chemicals
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™
CAS: 1730-25-2 | C3H5BrMg | 145.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA209531000
Price (MYR)
612.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
612.00
EA
Add to cart
Specifications
Chemical Name or MaterialAllylmagnesium bromide
Name Note1M solution in diethyl ether
CAS1730-25-2
CAS Min %15.37
Health Hazard 1GHS Signal Word: Danger