Trimethylsilyl trifluoromethanesulfonate, 99%
Trimethylsilyl trifluoromethanesulfonate, 99%
Trimethylsilyl trifluoromethanesulfonate, 99%
Trimethylsilyl trifluoromethanesulfonate, 99%
Thermo Scientific Chemicals
Trimethylsilyl trifluoromethanesulfonate, 99%
CAS: 27607-77-8 | C4H9F3O3SSi | 222.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
10 mL
50 mL
Packaging:
Glass bottle
Catalog number FSA209442500
Price (MYR)
3,652.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,652.00
EA
Add to cart
Specifications
Chemical Name or MaterialTrimethylsilyl trifluoromethanesulfonate
CAS27607-77-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Reacts violently with water.
Health Hazard 3GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.