3-Nitrobenzenesulfonic acid, sodium salt, 97%
3-Nitrobenzenesulfonic acid, sodium salt, 97%
3-Nitrobenzenesulfonic acid, sodium salt, 97%
3-Nitrobenzenesulfonic acid, sodium salt, 97%
Thermo Scientific Chemicals
3-Nitrobenzenesulfonic acid, sodium salt, 97%
CAS: 127-68-4 | C6H4NNaO5S | 225.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic Bottle
Catalog number FSA209120010
Price (MYR)
780.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
780.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Nitrobenzenesulfonic acid, sodium salt
CAS127-68-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0