4-(Trifluoromethyl)benzaldehyde, 98%
4-(Trifluoromethyl)benzaldehyde, 98%
4-(Trifluoromethyl)benzaldehyde, 98%
4-(Trifluoromethyl)benzaldehyde, 98%
Thermo Scientific Chemicals
4-(Trifluoromethyl)benzaldehyde, 98%
CAS: 455-19-6 | C8H5F3O | 174.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA208920500
Price (MYR)
1,472.00
BOT
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,472.00
BOT
Add to cart
Specifications
Chemical Name or Material4-(Trifluoromethyl)benzaldehyde
Name Note95%
CAS455-19-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning