Iodine monobromide, 98%
Iodine monobromide, 98%
Iodine monobromide, 98%
Iodine monobromide, 98%
Thermo Scientific Chemicals
Iodine monobromide, 98%
CAS: 7789-33-5 | BrI | 206.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number FSA208842500
Price (MYR)
1,645.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,645.00
EA
Add to cart
Specifications
Boiling Point116.0°C
Chemical Name or MaterialIodine monobromide
Melting Point42.0°C to 50.0°C
CAS7789-33-5
ColorBlack-Brown to Gray
View more
C-X bond formation reagent
RUO – Research Use Only