Bismuth(III) chloride, 97+%, anhydrous
Bismuth(III) chloride, 97+%, anhydrous
Bismuth(III) chloride, 97+%, anhydrous
Bismuth(III) chloride, 97+%, anhydrous
Thermo Scientific Chemicals
Bismuth(III) chloride, 97+%, anhydrous
CAS: 7787-60-2 | BiCl3 | 315.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA208835000
Price (MYR)
5,017.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,017.00
EA
Add to cart
Specifications
Boiling Point447.0°C
Chemical Name or MaterialBismuth(III) chloride
Melting Point230.0°C to 232.0°C
Name NoteAnhydrous
CAS7787-60-2