Ammonium dichromate, 99%
Ammonium dichromate, 99%
Ammonium dichromate, 99%
Ammonium dichromate, 99%
Thermo Scientific Chemicals
Ammonium dichromate, 99%
CAS: 7789-09-5 | H8Cr2N2O7 | 252.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA208815000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium dichromate
Melting Point170.0°C
Name Note99%
CAS7789-09-5
ColorOrange
View more
Oxidation reagent
RUO – Research Use Only