Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%
Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%
Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%
Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%
Thermo Scientific Chemicals
Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%
CAS: 50525-27-4 | C30H36Cl2N6O6Ru | 748.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA208750050
Price (MYR)
3,135.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,135.00
EA
Add to cart
Specifications
Chemical Name or MaterialTris(2, 2'-bipyridyl)ruthenium(II) chloride, hexahydrate
CAS50525-27-4
Melting Point>300.0°C
Elemental Analysis97.5% min.
Infrared SpectrumAuthentic