Succinic acid, disodium salt, 99%, anhydrous
Succinic acid, disodium salt, 99%, anhydrous
Succinic acid, disodium salt, 99%, anhydrous
Succinic acid, disodium salt, 99%, anhydrous
Thermo Scientific Chemicals
Succinic acid, disodium salt, 99%, anhydrous
CAS: 150-90-3 | C4H4Na2O4 | 162.052 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA208745000
Price (MYR)
405.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
405.00
EA
Add to cart
Specifications
Chemical Name or MaterialSuccinic acid, disodium salt
Name Note0.99
CAS150-90-3
Health Hazard 1HYGROSCOPIC
Melting Point>300.0°C