Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™
Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™
Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™
Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™
Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™
Thermo Scientific Chemicals
Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™
CAS: 32916-51-1 | C5H9ClMg | 128.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA208461000
Price (MYR)
542.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
542.00
EA
Add to cart
Specifications
Chemical Name or MaterialCyclopentylmagnesium chloride
Name Note2M solution in diethyl ether
CAS32916-51-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
Extremely flammable liquid and vapor.
May form explosive