4-Methylmorpholine N-oxide hydrate, 97%
4-Methylmorpholine N-oxide hydrate, 97%
4-Methylmorpholine N-oxide hydrate, 97%
4-Methylmorpholine N-oxide hydrate, 97%
4-Methylmorpholine N-oxide hydrate, 97%
Thermo Scientific Chemicals
4-Methylmorpholine N-oxide hydrate, 97%
CAS: 80913-66-2 | C5H11NO2 | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA208340250
Price (MYR)
473.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
473.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Methylmorpholine N-oxide hydrate
Name Note97%
CAS80913-66-2
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
View more
Oxidation reagent
RUO – Research Use Only