Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Thermo Scientific Chemicals
Phosphotungstic acid hydrate
CAS: 12501-23-4 | H5O41PW12 | 2880.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA208315000
Price (MYR)
3,431.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
3,431.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhosphotungstic acid hydrate
Name Notep.a.
CAS12501-23-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement: