Ethylene glycol diethyl ether, 99%
Ethylene glycol diethyl ether, 99%
Ethylene glycol diethyl ether, 99%
Ethylene glycol diethyl ether, 99%
Thermo Scientific Chemicals
Ethylene glycol diethyl ether, 99%
CAS: 629-14-1 | C6H14O2 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA208241000
Price (MYR)
396.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
396.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthylene glycol diethyl ether
CAS629-14-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye irritation.
May damage the unborn child.
Suspected of damaging fertility.
Highly flammable liquid and vapor.
May form explosive peroxides.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES