Sodium carbonate, 99.8%, for analysis, anhydrous, powder
Sodium carbonate, 99.8%, for analysis, anhydrous, powder
Sodium carbonate, 99.8%, for analysis, anhydrous, powder
Sodium carbonate, 99.8%, for analysis, anhydrous, powder
Thermo Scientific Chemicals
Sodium carbonate, 99.8%, for analysis, anhydrous, powder
CAS: 497-19-8 | CNa2O3 | 105.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic Bottle
Catalog number FSA207810010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Boiling Point1600.0°C
Chemical Name or MaterialSodium carbonate
Melting Point851.0°C
Name NoteAnhydrous
CAS497-19-8
View more
Adsorbent, Biological buffer
RUO – Research Use Only