Calcium chloride dihydrate, 99+%, for analysis
Calcium chloride dihydrate, 99+%, for analysis
Calcium chloride dihydrate, 99+%, for analysis
Calcium chloride dihydrate, 99+%, for analysis
Thermo Scientific Chemicals
Calcium chloride dihydrate, 99+%, for analysis
CAS: 10035-04-8 | CaCl2·2H2O | 147.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Plastic Bottle
Catalog number FSA207780025
Price (MYR)
1,388.00
EA
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,388.00
EA
Add to cart
Specifications
Chemical Name or MaterialCalcium chloride dihydrate
Melting Point175.0°C
CAS10035-04-8
ColorWhite
Health Hazard 1GHS Signal Word: Warning
View more
Adsorbent
RUO – Research Use Only