Potassium oxalate monohydrate, 99+%, for analysis
Potassium oxalate monohydrate, 99+%, for analysis
Potassium oxalate monohydrate, 99+%, for analysis
Potassium oxalate monohydrate, 99+%, for analysis
Thermo Scientific Chemicals
Potassium oxalate monohydrate, 99+%, for analysis
CAS: 6487-48-5 | C2H2K2O5 | 184.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA207715000
Price (MYR)
471.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
471.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPotassium oxalate monohydrate
Name Notep.a.
CAS6487-48-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement