Copper(II) nitrate trihydrate, 99%, for analysis
Copper(II) nitrate trihydrate, 99%, for analysis
Copper(II) nitrate trihydrate, 99%, for analysis
Copper(II) nitrate trihydrate, 99%, for analysis
Thermo Scientific Chemicals
Copper(II) nitrate trihydrate, 99%, for analysis
CAS: 10031-43-3 | CuN2O6·3H2O | 241.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA207685000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper(II) nitrate trihydrate
Melting Point114.0°C
Name Notep.a.
CAS10031-43-3
ColorBrown to Cream