Tri-n-octylphosphine oxide, 99%
Tri-n-octylphosphine oxide, 99%
Tri-n-octylphosphine oxide, 99%
Tri-n-octylphosphine oxide, 99%
Thermo Scientific Chemicals
Tri-n-octylphosphine oxide, 99%
CAS: 78-50-2 | C24H51OP | 386.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA207601000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTri-n-octylphosphine oxide
CAS78-50-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Suspected of damaging fertility or the unborn child.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protecti