gamma-Terpinene, 97%, stabilized
gamma-Terpinene, 97%, stabilized
gamma-Terpinene, 97%, stabilized
gamma-Terpinene, 97%, stabilized
Thermo Scientific Chemicals
gamma-Terpinene, 97%, stabilized
CAS: 99-85-4 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA207505000
Price (MYR)
1,250.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,250.00
EA
Add to cart
Specifications
Chemical Name or Materialγ-Terpinene
Name NoteStabilized
CAS99-85-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.