alpha-Terpinene, 90%, tech.
alpha-Terpinene, 90%, tech.
alpha-Terpinene, 90%, tech.
alpha-Terpinene, 90%, tech.
Thermo Scientific Chemicals
alpha-Terpinene, 90%, tech.
CAS: 99-86-5 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA207491000
Price (MYR)
289.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
289.00
BOT
Add to cart
Specifications
Chemical Name or Materialα-Terpinene
Name Notetech., 90%
CAS99-86-5
CAS Min %88.0
Health Hazard 1GHS Signal Word: Danger