Squalene, 99+%, Thermo Scientific Chemicals
Squalene, 99+%, Thermo Scientific Chemicals
Squalene, 99+%, Thermo Scientific Chemicals
Squalene, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Squalene, 99+%, Thermo Scientific Chemicals
Squalene, CAS # 111-02-4, is a biosynthetic precursor to all steroids and ergosterol and is used as an immunologic adjuvant in vaccines.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA207471000
Price (MYR)
664.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
664.00
EA
Add to cart
Specifications
Chemical Name or MaterialSqualene
Acidity1mg KOH/g max.
CAS111-02-4
ColorColorless to Yellow
Density0.855g/mL
View more
General description
• Squalene is a precursor to steroids and ergosterols
• It can be used in preparation of lipids, steroids, and fat-soluble vitamins
Applications
• Squalene is used as an immunologic adjuvant in vaccines
• It plays a vital role in the preparation of cholesterol, steroid hormones, and vitamin D
• It can also serve as a lubricant for fatty acid surfactant films adsorbed on iron oxide surfaces
RUO – Research Use Only
General References:
  1. Sánchez-Fidalgo, S.; Villegas, I.; Rosillo, M. Á.; Aparicio-Soto, M.; de la Lastra, C. A. Dietary squalene supplementation improves DSS-induced acute colitis by downregulating p38 MAPK and NFkB signaling pathways. Mol. Nutr. Food Res. 2015, 59 (2), 284-292.
  2. Kedl, J. D.; Kedl, R. M. How squalene GLAdly helps generate antigen-specific T cells via antigen-carrying neutrophils and IL-18. Eur. J. Immunol. 2015, 45 (2), 376-379.