all-trans-Retinoic acid, 97%
all-trans-Retinoic acid, 97%
all-trans-Retinoic acid, 97%
all-trans-Retinoic acid, 97%
Thermo Scientific Chemicals
all-trans-Retinoic acid, 97%
All-trans-retinol, Vitamin A, retinol, CAS # 68-26-8, is a fat-soluble vitamin that binds retinoid receptors within cells. | CAS: 302-79-4 | C20H28O2 | 300.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA207341000
Price (MYR)
440.00
BOT
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
440.00
BOT
Add to cart
Specifications
Chemical Name or Materialall-trans-Retinoic acid
Name Note97%
CAS302-79-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
View more
General description
• All-trans-retinol, commonly known as Vitamin A, is a fat-soluble essential nutrient
• All-trans-retinol is involved in hormonal signaling via the retinoid receptors
• It is an important regulator of cell division and apoptosis
• Deficiency can lead to vision, bone, immune, and skin disorders
Applications
• Suitable for research on cell differentiation, embryonic development, immunology and more
• Laboratory grade quality, for research use only
RUO – Research Use Only
General References:
  1. Stephensen, C.B.; Lietz, G. Vitamin A in resistance to and recovery from infection: relevance to SARS-CoV2. Br J Nutr. 2021, 126(11), 1663-1672.
  2. He, J.; Gu, Y.; Zhang, S. Vitamin A and Breast Cancer Survival: A Systematic Review and Meta-analysis. Clin Breast Cancer. 2018, 18(6), e1389-e1400.