1-Phenyl-2-thiourea, 97%
1-Phenyl-2-thiourea, 97%
1-Phenyl-2-thiourea, 97%
1-Phenyl-2-thiourea, 97%
Thermo Scientific Chemicals
1-Phenyl-2-thiourea, 97%
CAS: 103-85-5 | C7H8N2S | 152.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA207250250
Price (MYR)
601.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
601.00
EA
Add to cart
Specifications
Chemical Name or Material1-Phenyl-2-thiourea
CAS103-85-5
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0