Nonadecanoic acid, 98+%
Nonadecanoic acid, 98+%
Nonadecanoic acid, 98+%
Nonadecanoic acid, 98+%
Thermo Scientific Chemicals
Nonadecanoic acid, 98+%
CAS: 646-30-0 | C19H38O2 | 298.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA207190010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialNonadecanoic acid
CAS646-30-0
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0