5-Methylisatin, 95%
5-Methylisatin, 95%
5-Methylisatin, 95%
Thermo Scientific Chemicals
5-Methylisatin, 95%
CAS: 608-05-9 | C9H7NO2 | 161.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA206860010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Methylisatin
CAS608-05-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.