Vanadium(V) oxide, 99.6+%
Vanadium(V) oxide, 99.6+%
Vanadium(V) oxide, 99.6+%
Vanadium(V) oxide, 99.6+%
Thermo Scientific Chemicals
Vanadium(V) oxide, 99.6+%
CAS: 1314-62-1 | O5V2 | 181.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA206422500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point1750.0°C
Chemical Name or MaterialVanadium(V) oxide
Melting Point690.0°C
Name Note99.6+%
CAS1314-62-1