Copper(II) chloride dihydrate, 98%, pure
Copper(II) chloride dihydrate, 98%, pure
Copper(II) chloride dihydrate, 98%, pure
Copper(II) chloride dihydrate, 98%, pure
Thermo Scientific Chemicals
Copper(II) chloride dihydrate, 98%, pure
CAS: 10125-13-0 | Cl2CuH4O2 | 170.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA206345000
Price (MYR)
1,176.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,176.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCopper(II) chloride dihydrate
Melting Point100.0°C
Name NoteTechnical, 98%
CAS10125-13-0