Ammonium cerium(IV) sulfate dihydrate, 99+%, extra pure
Ammonium cerium(IV) sulfate dihydrate, 99+%, extra pure
Ammonium cerium(IV) sulfate dihydrate, 99+%, extra pure
Ammonium cerium(IV) sulfate dihydrate, 99+%, extra pure
Thermo Scientific Chemicals
Ammonium cerium(IV) sulfate dihydrate, 99+%, extra pure
CAS: 10378-47-9 | H16CeN4O16S4·2H2O | 632.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA206315000
Price (MYR)
2,324.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,324.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium cerium(IV) sulfate dihydrate
Melting Point130.0°C
Name NoteExtra Pure
CAS10378-47-9
ColorOrange