Boron oxide, 98%, -40 mesh
Boron oxide, 98%, -40 mesh
Boron oxide, 98%, -40 mesh
Boron oxide, 98%, -40 mesh
Boron oxide, 98%, -40 mesh
Thermo Scientific Chemicals
Boron oxide, 98%, -40 mesh
CAS: 1303-86-2 | B2O3 | 69.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Packaging:
View all
Plastic Drum
Catalog number FSA206300050
Price (MYR)
1,760.00
EA
Add to cart
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
1,760.00
EA
Add to cart
Specifications
Boiling Point1860.0°C
Chemical Name or MaterialBoron oxide
Melting Point450.0°C
CAS1303-86-2
ColorWhite
View more
Suitable for battery materials development.
RUO – Research Use Only