Ammonium oxalate monohydrate, 99.5%, for analysis
Ammonium oxalate monohydrate, 99.5%, for analysis
Ammonium oxalate monohydrate, 99.5%, for analysis
Ammonium oxalate monohydrate, 99.5%, for analysis
Thermo Scientific Chemicals
Ammonium oxalate monohydrate, 99.5%, for analysis
CAS: 6009-70-7 | C2H8N2O4·H2O | 142.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA206275000
Price (MYR)
521.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
521.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAmmonium oxalate monohydrate
Melting Point70.0°C
Name Notep.a.
CAS6009-70-7