Aluminum ammonium sulfate dodecahydrate, 99%, pure
Aluminum ammonium sulfate dodecahydrate, 99%, pure
Aluminum ammonium sulfate dodecahydrate, 99%, pure
Aluminum ammonium sulfate dodecahydrate, 99%, pure
Thermo Scientific Chemicals
Aluminum ammonium sulfate dodecahydrate, 99%, pure
CAS: 7784-26-1 | H4AlNO8S2·12H2O | 453.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number FSA206261000
Price (MYR)
287.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
287.00
EA
Add to cart
Specifications
Boiling Point200.0°C
Chemical Name or MaterialAluminum ammonium sulfate dodecahydrate
Melting Point93.5°C
CAS7784-26-1