1-hexanesulfonic acid, sodium salt hydrate, 99%
1-hexanesulfonic acid, sodium salt hydrate, 99%
1-hexanesulfonic acid, sodium salt hydrate, 99%
1-hexanesulfonic acid, sodium salt hydrate, 99%
Thermo Scientific Chemicals
1-hexanesulfonic acid, sodium salt hydrate, 99%
CAS: 2832-45-3 | C6H13NaO3S | 188.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA206121000
Price (MYR)
2,772.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,772.00
EA
Add to cart
Specifications
Chemical Name or Material1-Hexanesulfonic acid, sodium salt hydrate
CAS2832-45-3
Absorbance5.5 w/v% aq. soln., pH 3, H3PO4, 0.06 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm, 5.5 w/v% aq. soln., pH 3, H3PO4 at 0nm
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement