1-Butanesulfonic acid, sodium salt, 98%
1-Butanesulfonic acid, sodium salt, 98%
1-Butanesulfonic acid, sodium salt, 98%
1-Butanesulfonic acid, sodium salt, 98%
Thermo Scientific Chemicals
1-Butanesulfonic acid, sodium salt, 98%
CAS: 2386-54-1 | C4H9NaO3S | 160.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA206101000
Price (MYR)
2,314.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,314.00
EA
Add to cart
Specifications
Chemical Name or Material1-Butanesulfonic acid, sodium salt
CAS2386-54-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0