Sodium tetraborate decahydrate, 99.5%, for analysis
Sodium tetraborate decahydrate, 99.5%, for analysis
Sodium tetraborate decahydrate, 99.5%, for analysis
Sodium tetraborate decahydrate, 99.5%, for analysis
Thermo Scientific Chemicals
Sodium tetraborate decahydrate, 99.5%, for analysis
CAS: 1303-96-4 | B4Na2O7·10H2O | 381.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
1 kg
5 kg
Packaging:
View all
Plastic Drum
Catalog number FSA205950100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 kg
Packaging:
Plastic Drum
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium tetraborate decahydrate
Melting Point75.0°C
CAS1303-96-4
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
Biological buffer for analysis
RUO – Research Use Only