Manganese(II) sulfate monohydrate, 99+%, extra pure
Manganese(II) sulfate monohydrate, 99+%, extra pure
Manganese(II) sulfate monohydrate, 99+%, extra pure
Manganese(II) sulfate monohydrate, 99+%, extra pure
Manganese(II) sulfate monohydrate, 99+%, extra pure
Thermo Scientific Chemicals
Manganese(II) sulfate monohydrate, 99+%, extra pure
CAS: 10034-96-5 | H2MnO5S | 169.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA205905000
Price (MYR)
522.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
522.00
BOT
Add to cart
Specifications
Boiling Point850.0°C
Chemical Name or MaterialManganese(II) sulfate monohydrate
Melting Point700.0°C
CAS10034-96-5
ColorPink to White
View more
Suitable for battery materials development.
RUO – Research Use Only