Ammonium nitrate, 99+%, for analysis
Ammonium nitrate, 99+%, for analysis
Ammonium nitrate, 99+%, for analysis
Ammonium nitrate, 99+%, for analysis
Thermo Scientific Chemicals
Ammonium nitrate, 99+%, for analysis
Ammonium nitrate, 99+%, H4N2O3, CAS Number-6484-52-2, ammonium nitricum, ammonium saltpeter, merco prills, herco prills, nitric acid ammonium salt, nitram, nitrato amonico, nitrate of ammonia, german saltpeter, ammonium nitrate, 500g, 210.0 deg.C, CHEBI:63038, 5ppm max, 5ppm max., White, 1.7200g/mL | CAS: 6484-52-2 | H4N2O3 | 80.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA205865000
Price (MYR)
451.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
451.00
EA
Add to cart
Specifications
Boiling Point210.0°C
Chemical Name or MaterialAmmonium nitrate
Melting Point169.0°C
Name Notep.a.
CAS6484-52-2
View more
For analysis
RUO – Research Use Only