Formamide, 99.5%, for analysis
Formamide, 99.5%, for analysis
Formamide, 99.5%, for analysis
Formamide, 99.5%, for analysis
Thermo Scientific Chemicals
Formamide, 99.5%, for analysis
CAS: 75-12-7 | CH3NO | 45.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
Catalog number FSA205821000
Price (MYR)
350.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
350.00
EA
Add to cart
Specifications
Chemical Name or MaterialFormamide
Melting Point2.0°C to 3.0°C
CAS75-12-7
Health Hazard 1Danger
Health Hazard 2GHS P Statement May damage the unborn child. May cause damage to organs through prolonged or repeated exposure. Suspected of causing cancer if swallowed.
View more
For analysis
RUO – Research Use Only