Manganese(II) acetate tetrahydrate, 99+%, for analysis
Manganese(II) acetate tetrahydrate, 99+%, for analysis
Manganese(II) acetate tetrahydrate, 99+%, for analysis
Manganese(II) acetate tetrahydrate, 99+%, for analysis
Manganese(II) acetate tetrahydrate, 99+%, for analysis
Thermo Scientific Chemicals
Manganese(II) acetate tetrahydrate, 99+%, for analysis
CAS: 6156-78-1 | C4H14MnO8 | 245.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA205665000
Price (MYR)
367.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
367.00
BOT
Add to cart
Specifications
Chemical Name or MaterialManganese(II) acetate tetrahydrate
Melting Point>300.0°C
Name Note99+%
CAS6156-78-1
ColorPink
View more
Suitable for battery materials development.
RUO – Research Use Only