Florisil™, 60-100 mesh, for column chromatography
Florisil™, 60-100 mesh, for column chromatography
Florisil™, 60-100 mesh, for column chromatography
Florisil™, 60-100 mesh, for column chromatography
Thermo Scientific Chemicals
Florisil™, 60-100 mesh, for column chromatography
Florisil, MgO3Si, CAS Number-1343-88-0, 1kg, White, 01,393; 09,223, 100.39, ZADYMNAVLSWLEQ-UHFFFAOYSA-N, 10, 5509, MFCD00078249, 60-100 mesh, for column chromatography, Glass bottle | CAS: 1343-88-0 | MgO3Si | 100.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Catalog number FSA205455000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialFlorisil™
ColorWhite
Physical FormFine Powder
Name Note60-100 mesh, for column chromatography
CAS1343-88-0
View more
for column chromatography
RUO – Research Use Only