(S)-(-)-alpha-Methylbenzyl isocyanate, 98%
(S)-(-)-alpha-Methylbenzyl isocyanate, 98%
(S)-(-)-alpha-Methylbenzyl isocyanate, 98%
(S)-(-)-alpha-Methylbenzyl isocyanate, 98%
Thermo Scientific Chemicals
(S)-(-)-alpha-Methylbenzyl isocyanate, 98%
CAS: 14649-03-7 | C9H9NO | 147.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
Packaging:
Glass bottle
Catalog number FSA205310010
Price (MYR)
650.00
EA
Add to cart
Quantity:
1 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
650.00
EA
Add to cart
Specifications
Chemical Name or Material(S)-(-)-α-Methylbenzyl isocyanate
Name Note98%
CAS14649-03-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
View more
Chiral derivatization reagent
RUO – Research Use Only