Ethyl methanesulfonate, 99%
Ethyl methanesulfonate, 99%
Ethyl methanesulfonate, 99%
Ethyl methanesulfonate, 99%
Thermo Scientific Chemicals
Ethyl methanesulfonate, 99%
CAS: 62-50-0 | C3H8O3S | 124.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA205261000
Price (MYR)
2,491.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,491.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthyl methanesulfonate
CAS62-50-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
May cause genetic defects.
May cause cancer.
Causes skin irritation.
Harmful if swallowed.
Health Hazard 3GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.