Aluminum isopropoxide, 98%
Aluminum isopropoxide, 98%
Aluminum isopropoxide, 98%
Aluminum isopropoxide, 98%
Thermo Scientific Chemicals
Aluminum isopropoxide, 98%
CAS: 555-31-7 | C9H21AlO3 | 204.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA205175000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum isopropoxide
CAS555-31-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable solid.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing d
View more
Lewis acid
RUO – Research Use Only