Magnesium oxide, 98%, pure
Magnesium oxide, 98%, pure
Magnesium oxide, 98%, pure
Magnesium oxide, 98%, pure
Thermo Scientific Chemicals
Magnesium oxide, 98%, pure
CAS: 1309-48-4 | MgO | 40.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA205155000
Price (MYR)
360.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
360.00
BOT
Add to cart
Specifications
Boiling Point3600°C
Chemical Name or MaterialMagnesium oxide
Melting Point2800°C
Name Note98%, ca 40 mesh
CAS1309-48-4