DL-Isocitric acid, trisodium salt hydrate, 95%
DL-Isocitric acid, trisodium salt hydrate, 95%
DL-Isocitric acid, trisodium salt hydrate, 95%
DL-Isocitric acid, trisodium salt hydrate, 95%
DL-Isocitric acid, trisodium salt hydrate, 95%
DL-Isocitric acid, trisodium salt hydrate, 95%
Thermo Scientific Chemicals
DL-Isocitric acid, trisodium salt hydrate, 95%
CAS: 1637-73-6 | C6H5Na3O7 | 258.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA205010010
Price (MYR)
433.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
433.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDL-Isocitric acid, trisodium salt hydrate
CAS1637-73-6
ColorWhite
EINECS Number216-668-3
Infrared SpectrumAuthentic