Isethionic acid, sodium salt, 98%
Isethionic acid, sodium salt, 98%
Isethionic acid, sodium salt, 98%
Isethionic acid, sodium salt, 98%
Thermo Scientific Chemicals
Isethionic acid, sodium salt, 98%
CAS: 1562-00-1 | C2H5NaO4S | 148.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA205005000
Price (MYR)
460.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
460.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIsethionic acid, sodium salt
Name Note98%
CAS1562-00-1
Health Hazard 1HYGROSCOPIC
Melting Point191.0°C to 194.0°C